Learn More
TDRD3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$155.13 - $588.46
Specifications
Antigen | TDRD3 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2628
![]() |
Bethyl Laboratories, Inc
IHC00737T |
10 μL |
Each for $155.13
|
|
|||||
50-157-2627
![]() |
Bethyl Laboratories, Inc
IHC00737 |
100 μL |
Each for $588.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Rabbit, Pig, Panda, Orangutan
TDRD3 belongs to the tudor domain group of proteins, which are prevalent among proteins involved in RNA metabolism. TDRD3 is widely expressed and localizes predomitly in the cytoplasm. It is detected in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas. Overexpression of TDRD3 in cells induces the formation of stress granules. Multiple isoforms of TDRD3 exist due to alternative splicing varients. In humans, the gene is located on chromosome 13.Specifications
TDRD3 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Tudor domain-containing protein 3 | |
TDRD3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9H7E2 | |
81550 | |
Between 601 and 651 | |
Primary | |
4° C | |
TDRD3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.