Learn More
TdT Mouse anti-Human, FITC, Clone: TdT-6, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHTDT015
 View more versions of this product
Catalog No. 50-112-1486
Add to cart
Description
Description
Based on our testing, publications, and results reported from customers, the Alexa Fluor™ 488 dye provides the best fluorescein (FITC) substitute.
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3 ft. hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt ft.s lymphoma.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
TdT
Monoclonal
FITC
DNTT
Liquid
DNTT
Human Terminal Deoxynucleotidyl Transferase (TdT)
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
TdT-6
PBS with 1% BSA and 0.1% sodium azide
P04053
TDT, DNA nucleotidylexotransferase, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, terminal addition enzyme, terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, terminal transferase
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
1791