Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TdT Mouse anti-Human, Mouse, Rat, Clone: 7UNA8U6, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14973980

 View more versions of this product

Catalog No. 50-112-2805


Add to cart

Description

Description

Description: The monoclonal antibody 7UNA8U6 reacts with human, mouse, and rat TdT. TdT, or terminal-deoxynucleotidyl transferase, is a 60 kDa polymerase responsible for the template-independent addition of N-nucleotides at gene segment junctions of developing lymphocytes. Expression is found in the nucleus of immature lymphocytes but not in mature lymphocytes or non-lymphoid cells. TdT plays a critical role during TCR and Ig gene rearrangement and functions to increase the diversity of the lymphocyte repertoire through the addition of N-nucleotides onto the ends of newly formed P-nucleotides. After positive selection, expression of TdT becomes decreased or absent. Expression of TdT has been shown to be present in lymphoma or lymphoblastic leukemia cells. Applications Reported: This 7UNA8U6 antibody has been reported for use in western blotting and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections. Applications Tested: This 7UNA8U6 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human and mouse tissue using low pH antigen retrieval and can be used at less than or equal to 10 µg/mL. This 7UNA8U6 antibody has also been tested by western blot analysis of reduced cell lysate prepared from mouse and rat cells and can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3 ft. hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt ft.s lymphoma.
Specifications

Specifications

TdT
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P04053, P09838
TDT
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
7UNA8U6
Unconjugated
DNTT
Liquid
DNTT
IgG1, kappa
25 μg
4° C
Primary
1791, 21673, 294051
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.