Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TdT Rabbit anti-Human, Clone: DNTT/4506R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen TdT
Clone DNTT/4506R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122842
View Documents
Novus Biologicals
NBP30768420UG
20 μg
Each for $159.82
Add to cart
 
NB122841
View Documents
Novus Biologicals
NBP30768410UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

TdT Monoclonal antibody specifically detects TdT in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TdT
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase
Synthetic peptide corresponding to residues within aa1-100 of TdT was used as an immunogen.
Monoclonal
Protein A or G purified
DNTT/4506R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Adaptive Immunity, Cell Cycle and Replication, Immunology
10 mM PBS with 0.05% BSA
1791
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit