Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse Monoclonal Antibody

TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse MAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Manufacturer:  Thermo Scientific™ MS1105R7

 View more versions of this product

Catalog No. MS1105R7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1, Mouse Monoclonal Antibody.

Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.

Host Species: Mouse

Clone: SEN28

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: N-terminal

Immunogen: Recombinant protein corresponding to the amino terminal region of the TdT molecule

Molecular Weight: 58kDa

Positive Control: Thymus

Cellular Localization: Nuclear Ab-1 will be of use in the differentiation of lymphoblastic lymphoma from other lymphomas.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

TdT/Terminal Deoxyribonucleotidyl Transferase Ab-1
Monoclonal
Unconjugated
Murine
IgG2a
IVD
Store at 4°C, Do Not Freeze
Primary
Human
Immunohistochemistry (Paraffin)
SEN28
Prediluted
Recombinant protein corresponding to the amino terminal region of the TdT molecule
7mL
DNA Repair
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit