Learn More
Cytiva TE 62 Transfer Cooled Unit
TE 62 Transfer Cooled Unit - Load cassettes easily with even contact between the gel and transfer membrane.
Supplier: Cytiva TE62
This item is not returnable.
View return policy
Description
- TE 62 Transfer Cooled Unit
- New hinged cassettes molded of tough, glass-filled polyethersulfone for increased durability.
- Transfer four standard gels or 16 mini-gels.
- Load cassettes easily with even contact between the gel and transfer membrane.
- Secure, sturdy electrode panels lock automatically in place.
- Efficient transfer due to evenly spaced vertical platinum wire on electrode panel.
- The gel cassettes provide a stiff backbone for the gel and membrane sandwiches.
- The hinged halves lock together for equal pressure across the gel.
- The grid design of the gel cassettes maximizes the open area for even transfers of gels up to 15 x 21 cm.
- The crucial element for uniform electrophoretic transfer, a uniform electrical field, is assured by spacing the electrode panels 10 cm apart and incorporating 140 cm of wire in an evenly-spaced pattern.
- For safety, the electrode panels are locked securely into the Transfer Unit, but can be easily removed for cleaning or replacement.
Specifications
Safety Lid with Power Cables, 4 Gel Cassettes, 8 Foam Sponges 3 mm Thick, 4 Foam Sponges 6 mm Thick, 25 Sheets of Blotter Paper | |
Transfer Cooled Unit | |
4°C to 40°C | |
32 cm | |
16.5 cm |
TE 62 | |
1 Ea. | |
32 x 28 x 16.5 cm | |
28 cm |