Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Thermo Scientific™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Nalgene bottle
1185-53-1
C4H12ClNO3
157.594
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl
1185-53-1
C4H12ClNO3
157.594
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl

Use directly to re-suspend and/or dilute purified DNA or RNA

Concentration 1X
Physical Form Solution
Grade Molecular Biology
Packaging Nalgene bottle
pH 8
Quantity 100mL
Chemical Name or Material TE Buffer