Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TE buffer, (20X), pH 7.6, autoclaved, Alfa Aesar™

Click to view available options
Quantity:
250mL
500mL
1185-53-1
C4H12ClNO3
157.594
MFCD00236359
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl
1185-53-1
C4H12ClNO3
157.594
MFCD00236359
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
C(C(CO)(CO)N)O.Cl
Physical Form Liquid
Packaging Plastic bottle
Quantity 250mL
Chemical Name or Material TE buffer