Tech Glass Replacement Squeeze-O-Matic Burets with Glass Plugs
Replacement buret with Glass plug
Manufacturer: Tech Glass, Inc. TG184101016
 View more versions of this product
Catalog No. 10-436-35
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 
    Specifications
    Capacity (Metric) 10mL
    For Use With (Application) Used with acidic solutions
    For Use With (Equipment) 500mL LDPE bottle
    • Automatic self-zeroing
    • Used with low density polyethylene bottle
    • Used with acidic solutions
  • Description & Specifications
Related Products
Product Content Feedback