Learn More
Techne™ Filter Cartridges for PrimeQ Real-time PCR System
Incorporates both excitation and emission filters together with dichroic mirror for fluorophore excitation and emission detection
 View more versions of this product
Catalog No. 13-245-148
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Optimized for use with FAM™, HEX™, ROX™ and Cy™5 dyes
  • Fluorescence excitation range : 470-650nm (standard filters are centered at 485, 530, 580 and 640nm)
  • Fluorescence detection range : 500-710nm (standard filters are centered at 520, 560, 615 and 685nm)
  • FC02, FC03, FC04 and FC05 filter cartridges are supplied as standard with PrimeQ system
  • FC01 is not included with unit
Specifications
Specifications
Filter Cartridge
500λ
FAM multiplex, green intercalating dyes, Fluorescein, EvaGreen
460λ