Learn More
Techne™ Filter Cartridges for PrimeQ Real-time PCR System
Incorporates both excitation and emission filters together with dichroic mirror for fluorophore excitation and emission detection
Specifications
Kit Type Filter Cartridge
View More Specs
Products 1
Catalog Number Dye Type Excitation Value Wavelength Price Quantity & Availability  
Catalog Number Dye Type Excitation Value Wavelength Price Quantity & Availability  
13-245-148
View Documents Promotion Details
FAM multiplex, green intercalating dyes, Fluorescein, EvaGreen 460λ 500λ This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Optimized for use with FAM™, HEX™, ROX™ and Cy™5 dyes
  • Fluorescence excitation range : 470-650nm (standard filters are centered at 485, 530, 580 and 640nm)
  • Fluorescence detection range : 500-710nm (standard filters are centered at 520, 560, 615 and 685nm)
  • FC02, FC03, FC04 and FC05 filter cartridges are supplied as standard with PrimeQ system
  • FC01 is not included with unit
Specifications
Specifications
Filter Cartridge