Learn More
Techne™ Hybridization Incubator Accessory, Tubes
Versatile multiple tube formats; mix and match sizes within an instrument to cater for different throughputs, users and applications. Glass hybridization tubes reduce volumes, simplify washing and improve signals
 View more versions of this product
Catalog No. 22-312500
Add to cart
Description
Description
  • Tubes and other accessories can be accommodated at the same time for multiple uses
  • Low probe volume, even with large glass tubes
  • Rotation and design ensure volumes as low as 5ml can be used and recovered
  • Unique slot-in tubes with tube rotation speed of 0 to 20rpm are controllable to suit your application
  • Radioactive safe (protective casing and non-drip tube design minimizes risk to the user)
  • Thick borosilicate glass provides durability
  • Non-drip, safe and convenient easy pour design
  • O-ring sealed end caps ensure no leakage
  • Escape thread-on screw cap offer easy-opening with no vacuum problems
  • Tube assemblies feature adaptors to hold multiple small glass tubes for increased capacity
Specifications
Specifications
44mm
8.66 in.
Screw
Hybridization Tube
HB-1D Hybridizer
Borosilicate Glass
220mm