Techne™ Hybridization Incubator Accessory, Tubes

Versatile multiple tube formats; mix and match sizes within an instrument to cater for different throughputs, users and applications. Glass hybridization tubes reduce volumes, simplify washing and improve signals

 View more versions of this product

Catalog No. 13-245-115

  Check Availability
Add to Cart

    Specifications

    Diameter (Metric) Outer 80mm
    For Use With (Equipment) HB-1D Hybridizer
    Length (English) 9.44 in.
    Material Borosilicate Glass
    Closure Type Screw
    Length (Metric) 240mm
    Product Type Hybridization Tube

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit