Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TechniGlove Techni-Cleaned™ Gloves

Proprietary Microtexture Formers offer exceptional fit and a soft, supple feel.

Manufacturer:  Techniglove International IncSupplier Diversity Partner TN1203B

 View more versions of this product

Catalog No. 19-051-226C


  Check Availability
Add to cart

Specifications

Description

  • Manufactured in 3 colors
  • Meets requirements for particles and extractables
  • Contains no natural rubber or silicone
Description & Specifications

Specifications

Class 10
Beaded/Rolled
Nitrile
5 mil
Blue
12 in.
Micro textured
Ambidextrous
Large
Powder-free
Non-sterile
30.48cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit