Learn More
Tecniplast™ Tecniplast™ Accessories for Deluxe Reusable Animal Cages: Filter Covers
Manufacturer:  Tecniplast™ 1290D400SU
 View more versions of this product
Catalog No. 01-288-23C
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Filter Covers
  • For an isolated environment
  • Minimize temperature variations, humidity, harsh drafts, CO2 buildup
  • Promote animal health, encourage breeding; economical polyvinyl chloride (PVC) covers are disposable
  • Sturdy, molded polycarbonate (PC) covers are reusable
  • Use with disposable polyester fibers
Wire Lids
  • Extra strong
  • Crosswires prevent bending
  • 01-288-5H wires spaced 19mm apart; all others 7mm
  • Folding divider allows lid to be nested for storage
  • Autoclavable
  • Include protective stopper guard
Specifications
Specifications
Size Da/Db cages (16.75 x 10.5 x 6 in.)
Filter animal cage cover
Polysulfone, Stainless Steel