Learn More
Tecniplast™ Activity Wheel Cage System for Rodents
Ideal for research involving circadian rhythms, ingestion, drug or pharmaceutical testing
Manufacturer:  Tecniplast™ 2154F0105
Catalog No. 01-286-12C
Due to product restrictions, please to purchase or view availability for this product.
Includes: Activity wheel, wheel hub and support, sheet lid that encloses wheel and covers cage for secure containment, activity wire lid with water bottle support and food hopper
Description
Description
Allows easy, convenient measurement of lab rodents' voluntary physical activity in response to chemical or environmental stimuli.
  • Constructed with low-friction bushing for long life and quiet, smooth action All metal parts are stainless steel
  • All nonelectrical cage parts are autoclavable
  • Compatible with cages of dimensions 19 x 10.5 x 8 in. (48 x 27 x 20cm) (01-288-1F and 01-286-13F for rats); not included
Order Info
Accessory cage support (01-281-12B) and magnetic switch counter with LCD counter (01-286-12E) sold separately.
Specifications
Specifications
Yes
18.8 in.
Stainless Steel
31.5cm
Allows convenient measurement of lab Rodents' voluntary physical activity in response to chemical or environmental stimuli, Ideal for research involving circadian rhythms, ingestion, drug or pharmaceutical testing
48cm
12.4 in.
Activity wheel system
Documents
Accessories