Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tecniplast™ Disposable Polystyrene Animal Cages

For research involving radioactive, biohazardous, or toxic chemicals

Manufacturer:  Tecniplast™ 2154F005X

 View more versions of this product

Catalog No. 01-287-5B

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Dimensions (L x W x H): 48 x 27 x 20cm (19 x 10.5 x 8 in.)
  • Can be disposed of by incineration
  • Not autoclavable
  • Disposable
  • Polystyrene

Specifications

Specifications

10.5 in.
No
19 x 10.5 x 8 in. (48.2 x 26.6 x 20.32cm)
For research involving radioactive, biohazardous, or toxic chemicals
Polystyrene
8 in.
19 in.
Animal Cage
26.6cm
NRC
Yes
Mice, Rats, Hamsters, Guinea Pigs and Gerbils
942
20.32cm
48.2cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit