Learn More
Tecniplast™ Disposable Polystyrene Animal Cages
For research involving radioactive, biohazardous, or toxic chemicals
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
01-287-5B
View Documents
Tecniplast™
2154F005X
N/A 19 L x 10-1/2 W x 8 in. H
Each for $29.95
Case of 5 EA for $149.40
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Dimensions (L x W x H): 48 x 27 x 20cm (19 x 10.5 x 8 in.)
  • Can be disposed of by incineration
  • Not autoclavable
  • Disposable
  • Polystyrene