Learn More
Tecniplast™ Disposable Polystyrene Animal Cages
For research involving radioactive, biohazardous, or toxic chemicals
Manufacturer:  Tecniplast™ 1264C005
 View more versions of this product
Catalog No. 01-287-5A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Dimensions (L x W x H): 48 x 27 x 20cm (19 x 10.5 x 8 in.)
  • Can be disposed of by incineration
  • Not autoclavable
  • Disposable
  • Polystyrene