Learn More
Tecniplast™ Stainless Steel Raised Cage Lids without Divider
Secure and easty to handle
Specifications
Product Type Raised cage lid without divider
For Use With (Application) Guinea Pigs
Material Stainless steel
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
01-288-5K View Documents Tecniplast™
2000P242
Cage size P Each for $263.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • U-shaped hopper allows easy access to food and won't entrap food or contaminants
  • Cross-wire members prevent bending
  • 0.19cm wire spacing
  • Autoclavable
Specifications
Specifications
Raised cage lid without divider
Guinea Pigs
Stainless steel