Learn More
Tecniplast™ Tecniplast™ Stainless Steel Wire Cage Lids and Feeders Available on GSA/VA Contract for Federal Government customers only.
Secure and easty to handle
Manufacturer:  Tecniplast™ 2150E114
 View more versions of this product
Catalog No. 01-288-5E
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • U-shaped hopper allows easy access to food and won't entrap food or contaminants
  • Cross-wire members prevent bending
  • 0.07cm wire spacing
  • Folding hopper divider allows lids to be stacked for storage
  • Central divider prevents animal from crawling into feeder
  • Can be used with or without a raised wire floor
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Rats and Hamsters
Cage size E
Stainless Steel
0.07cm
Wire cage lid with feeder