Tecniplast™ Stainless Steel Wire Cage Lids and Feeders
Secure and easty to handle
Manufacturer: Tecniplast USA, IncSupplier Diversity Partner 1177A114
 View more versions of this product
Catalog No. 22-152116
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Material Stainelss steel
    For Use With (Equipment) Cage size B
    Product Type Wire cage lid with feeder
    Spacing (Metric) 0.07cm
    For Use With (Application) Mice
    • U-shaped hopper allows easy access to food and won't entrap food or contaminants
    • Cross-wire members prevent bending
    • 0.07cm wire spacing
    • Folding hopper divider allows lids to be stacked for storage
    • Central divider prevents animal from crawling into feeder
    • Can be used with or without a raised wire floor
    • Autoclavable
  • Description & Specifications
Related Products
Product Content Feedback