Learn More
Tecniplast™ ULTEM™ Animal Cages
Specifications
Autoclavable Autoclavable
Certifications/Compliance EU, USA and UK guidelines
Disposable No
View More Specs
Products 1
Catalog Number Mfr. No. Dimensions (L x W x H) Width (English) Surface Area (English) Material Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Width (English) Surface Area (English) Material Price Quantity & Availability  
02-934-122
View Documents
Tecniplast™
2000P001
61 x 43 x 20 cm 16.92 in. 320.07 sq. in. Polycarbonate
Each for $198.60
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
Autoclavable
EU, USA and UK guidelines
No