Learn More
Ted Pella Inc Carbon Tape, 20mm W x 20m L
Supplier: Ted Pella Inc 160732
These carbon tapes are conductive and may be used conveniently for scanning electron microscopy or EDS applications. This is the same material as the PELCO Tabs™, Carbon Conductive Tabs, but in tape form. The tape form allows for special application uses such as custom sizes, cut-outs and shapes. The purity of this tape is identical to the PELCO Tabs™; only very small impurities of Al and Si may be found. Both sides of the tape have a thick conductive adhesive (conductive inner film is 35µm and the adhesive is 45µm on each side for a total of 125µm [5 mils]) with a white liner. The double adhesive and conductive design permits quick mounting of samples without using liquid or colloidal adhesives.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.