Learn More
Cole-Parmer Parker Hannifin Parker Threaded Tee, PFA, 3/8" female NPT Fitting  DFS Item
Manufacturer:  Cole-Parmer 3132095
Threaded fittings are designed for more permanent installations. Choose the material and size that meets your application needs.
Catalog No. 50-632-046