Learn More
Cole-Parmer Parker Hannifin Compression union tee, PFA, 1/2"  DFS Item
Manufacturer:  Cole-Parmer 3132032
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. 50-631-996