Learn More
Cole-Parmer Parker Hannifin GTC-12 Compression union tee, PFA, 3/4"  DFS Item
Manufacturer:  Cole-Parmer 3132033
Able to withstand higher pressures than barbed fittings, compression fittings are an excellent choice to connect rigid tubing systems.
Catalog No. 50-631-992