Learn More
AVANTOR MASTERFLEX (VWR LLC) Parker Hannifin FTC-4 Flare union Tee, 1/4"", PFA, Each
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3049080
PFA wetted parts and PVDF nuts provide high purity and excellent chemical resistance. Leaf-free design withstands pressures to 225 psi. Ideal for high-vibration or side-load stressed applications. Low dead volume reduces particle entrapment, limits bacterial growth in deionized water, and simplifies rinsing.