Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TeenPrep™ Lysing Matrix J Available on GSA/VA Contract for Federal Government customers only.

Can be used with the TeenPrep. Certified RNaseand DNase free.

$86.50 - $846.50

Specifications

Product Type Lysing Matrix J
For Use With (Equipment) FastPrep-24, FastPrep 120; FastPrep-96
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP116936005 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MP Biomedicals™
116936005
5 x 15mL Each for $86.50
Add to cart
 
MP116936050 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MP Biomedicals™
116936050
50 x 15mL Each for $846.50
Add to cart
 
MP116936025 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MP Biomedicals™
116936025
25 x 15mL Each for $404.00
Add to cart
 
Description

Description

  • 15mL Lysing Matrix Tube pre-filled with Matrix-J
  • Can be used with the TeenPrep™ and the CoolTeenPrep™ adapters

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Lysing Matrix J
FastPrep-24, FastPrep 120; FastPrep-96
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit