Learn More
Cole-Parmer Masterflex Fitting, PVDF, Tee, Hose Barb to Threaded Adapter, 1/4" ID x 1/4" ID x 1/4" NPT(M); 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3070463
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
Catalog No. 50-630-570