Learn More
TEF1 Mouse anti-Human, DyLight 405, Clone: 13112, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243581V
Catalog No. NBP243581V
Add to cart
Description
Description
TEF1 Monoclonal antibody specifically detects TEF1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
TEF1
13112
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
TEAD1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human TEF1. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
NTEF-1, protein GT-IIC, TEA domain family member 1 (SV40 transcriptional enhancer factor), TEAD-1
Mouse
IgG
0.1 ml
Primary
7003