Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Teflon™ Bushing
GREENER_CHOICESDP

Teflon™ Bushing includes an EP™ O-ring, for use with pressure valves.

$30.78 - $44.46

Specifications

Autoclavable Not Autoclavable
Color White
Material PTFE
Shape Rounded Grip, Screw Stem
Sterility Non-sterile
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
31-502-262
GREENER_CHOICESDP
View Documents
Synthware™
B100015
15 mm
Each for $30.78
Only null left
Add to Cart
 
31-502-263
GREENER_CHOICESDP
View Documents
Synthware™
B100025
25 mm
Each for $44.46
Only null left
Add to Cart
 
Description

Description

  • For use with pressure valves
  • Includes an EP™ O-ring
Specifications

Specifications

Not Autoclavable
PTFE
Non-sterile
White
Rounded Grip, Screw Stem
Pressure Valves
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.