Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tegaserod maleate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
189188-57-6
C20H27N5O5
417.466
KIJRKSHEIAFGBH-BTJKTKAUSA-N
tegaserod maleate,sdz-htf 919,3-5-methoxy-1h-indol-3-ylmethylene-n-pentylcarbazimidamide hydrogen maleate,2-5-methoxy-1h-indol-3-yl methylene-n-pentylhydrazinecarboximidamide 2z-but-2-enedioic acid
57149572
(Z)-but-2-enedioic acid;1-[(5-methoxyindol-3-ylidene)methylamino]-2-pentylguanidine
CCCCCN=C(N)NNC=C1C=NC2=C1C=C(C=C2)OC.C(=CC(=O)O)C(=O)O
This item is not returnable. View return policy
189188-57-6
C20H27N5O5
417.466
KIJRKSHEIAFGBH-BTJKTKAUSA-N
tegaserod maleate,sdz-htf 919,3-5-methoxy-1h-indol-3-ylmethylene-n-pentylcarbazimidamide hydrogen maleate,2-5-methoxy-1h-indol-3-yl methylene-n-pentylhydrazinecarboximidamide 2z-but-2-enedioic acid
57149572
(Z)-but-2-enedioic acid;1-[(5-methoxyindol-3-ylidene)methylamino]-2-pentylguanidine
CCCCCN=C(N)NNC=C1C=NC2=C1C=C(C=C2)OC.C(=CC(=O)O)C(=O)O

5-HT4 partial agonist

Partial agonist of 5-HT4 (Ki = 12 nM).
Quantity 50mg
Formula Weight 417.46
Percent Purity >99%
Chemical Name or Material Tegaserod maleate

Recommended Storage : Store at +4°C