Learn More
TEKT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 139691AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-172-9128
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predomitly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis.
Specifications
Specifications
TEKT1
Polyclonal
Unconjugated
TEKT1
TEKT1
Rabbit
Antigen Affinity Chromatography
RUO
21689, 83659, 85270
-20°C
Liquid
Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q969V4, Q99JD2, Q9DAJ2
Tekt1
TEKT1 Fusion Protein Ag4993
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG