Learn More
TEKT2 Mouse anti-Human, Clone: OTI1E11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809970
Catalog No. 50-166-9820
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.
Specifications
Specifications
TEKT2
Monoclonal
Unconjugated
TEKT2
Lyophilized
TEKT2
Human recombit protein fragment corresponding to amino acids 1-244 of human TEKT2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1E11
PBS with 8% trehalose and no preservative; pH 7.3
Q9UIF3
TEKTB1, TEKTIN-T, h-tektin-t
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
27285