Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEKT2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549186
Description
Predicted to react with bovine, monkey and rat based on sequence homology.
Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.Specifications
TEKT2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UIF3 | |
TEKT2 | |
KLH conjugated synthetic peptide between 103-132 amino acids from the N-terminal region of human TEKT2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TEKT2 | |
Tektin-2, Tektin-t, Testicular tektin, Testicular tektin B1-like protein, TEKTB1, Tektin-B1, TEKT2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27285 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction