Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEKT4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | TEKT4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157589
|
Novus Biologicals
NBP157589 |
100 μL |
Each of 1 for $436.00
|
|
Description
TEKT4 Polyclonal specifically detects TEKT4 in Human samples. It is validated for Western Blot.Specifications
TEKT4 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
MGC27019, tektin 4, tektin-4 | |
TEKT4 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q8WW24 | |
150483 | |
Synthetic peptides corresponding to TEKT4(tektin 4) The peptide sequence was selected from the N terminal of TEKT4. Peptide sequence TSKYLLEEWFQNCYARYHQAFADRDQSERQRHESQQLATETQALAQRTQQ. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title