Learn More
Tele-Lite™ Honda™ BF135 Large Outboard Motors
Deliver advanced technology for a maximum boating experience
Manufacturer:  Tele-Lite™ TLBF135AKL
 View more versions of this product
Catalog No. 19-220-425
Add to cart
Description
Description
Provide an unmatched combination of power, torque, RPM and fuel efficiency.
  • 4-Stroke engine system
  • Water-cooled cooling system
  • Electric start
  • 5000-6000rpm
Specifications
Specifications
51A
2354cc
2.14:1 Gear ratio, F-N-R gear shift
Through hub
20 in.
50.8cm
3.4 in.
5000 to 6000rpm
8.7cm
478 lb.
217kg
2156.4 BTU/h
Water cooled
SOHC 4-stroke 4 cylinders/16 valves
Provide an unmatched combination of power, torque, rpm and fuel efficiency
135hp at 5500rpm
Microcomputer programmed
BF135, L-type
3.9 in.
Electric
9.9cm
22.8 in.
58cm
632w