Learn More
Tele Lite™ Honda™ BF135 Large Outboard Motors
Deliver advanced technology for a maximum boating experience
Manufacturer:  Tele-Lite, Inc.Supplier Diversity Partner TLBF135AKXC
 View more versions of this product
Catalog No. 19-220-427
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
51A
2354cc
2.14:1 Gear ratio, F-N-R gear shift
Through hub
25 in.
63.5cm
3.4 in.
5000 to 6000rpm
8.7cm
485 lb.
220kg
2156.4 BTU/h
Water cooled
SOHC 4-stroke 4 cylinders/16 valves
Provide an unmatched combination of power, torque, rpm and fuel efficiency
135hp at 5500rpm
Microcomputer programmed
BF135 (counter rotation), X-Type
3.9 in.
Electric
9.9cm
22.8 in.
58cm
632w