Tele Lite Lamp Head Assemblies
Manufacturer: Tele-Lite, Inc.Supplier Diversity Partner REP(2)5TLQ
 View more versions of this product
Catalog No. 19-130-1368
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product.
Please contact customer service for assistance: 1-800-766-7000.
Add to Cart
    Specifications
    For Use With (Equipment) For lamp
    Includes Outfitted with rough service bulb, heat tempered glass lens
    Voltage 120V
    Wattage Double 500w
    Product Type Lamp Head Assembly
  • Description & Specifications
Related Products
Product Content Feedback