Learn More
Tele-Lite Mister Rehab Adapter Kits
Use water mist to reduce ambient air temperature
Manufacturer: Tele-Lite TLAMK20
Description
Designed for personnel rehabilitation in hot environments. Adapter directs water vapor mist into discharge of smoke ejector fan. Resulting evaporation of mist reduces ambient air temperature as much as 10 to 20°.
- Connect to virtually any fan with spring clips
- Use less than two gallons of water per hour (require minimum 50psi water pressure)
- Connect to standard 0.75 in. dia. garden hose
- 6.75 in. deep and available in five sizes to fit various fan diameters