Learn More
Tele-Lite™ Mister Rehab Adapter Kits
Use water mist to reduce ambient air temperature
Products 5
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-220-319
View Documents
Tele-Lite™
TLAMK12
N/A Mister Kit 12 in. Fan This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-220-320
View Documents
Tele-Lite™
TLAMK16
N/A Mister Kit 16 in. Fan This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-220-321
View Documents
Tele-Lite™
TLAMK18
N/A Mister Kit 18 in. Fan This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-220-323
View Documents
Tele-Lite™
TLAMK24
N/A Mister Kit 21 in. Fan This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-220-322
View Documents
Tele-Lite™
TLAMK20
N/A Mister Kit 20 in. Fan This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Designed for personnel rehabilitation in hot environments. Adapter directs water vapor mist into discharge of smoke ejector fan. Resulting evaporation of mist reduces ambient air temperature as much as 10 to 20°.
  • Connect to virtually any fan with spring clips
  • Use less than two gallons of water per hour (require minimum 50psi water pressure)
  • Connect to standard 0.75 in. dia. garden hose
  • 6.75 in. deep and available in five sizes to fit various fan diameters