Learn More
Tele-Lite™ Gasoline-Powered Positive Pressure Fans
Gas-powered models have a direct-drive design allows for a more compact, lighter-weight package. Ideal for replacing a hostile interior environment with fresh, ambient air.
$3260.00 - $3720.00
Specifications
Product Type Positive Pressure Fan
Material Tubular steel
Color Red
View More Specs
Products 3
Catalog Number Mfr. No. Flow Rate Diameter (English) Fan Horsepower Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Flow Rate Diameter (English) Fan Horsepower Dimensions (L x W x H) Price Quantity & Availability  
19-220-301
View Documents
Tele-Lite™
TLAP21
19,900 cfm 21 in. 6.5 26.5ᅠxᅠ23 xᅠ27 in.
Each for $3,440.00
Add to cart
19-220-300
View Documents
Tele-Lite™
TLAP18
14,773 cfm 18 in. 5.5 22.5 x 23.5 x 23 in.
Each for $3,260.00
Add to cart
19-220-303
View Documents
Tele-Lite™
TLAP246
19,606 cfm 24 in. 6.5 28.25 x 22.5 x 30 in.
Each for $3,720.00
Add to cart
Description
Description
  • Powered by Honda™ engines
  • Quick setup requiring no power cords or generators
  • Side exhaust outlet and Oil Alert
  • Tubular steel frame
Specifications
Specifications
Positive Pressure Fan
Tubular steel
Red