Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tellurium ICP Standard, 10,000 ppm Te in 10% HNO3, Ricca Chemical
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
Specifications
Packaging | Natural LDPE Bottle |
Quantity | 100 mL |
CAS Min % | 67.0, 30.24, 0.67 |
CAS Max % | 70.0, 31.48, 0.7 |
Color | Colorless |
Concentration or Composition (by Analyte or Components) | 1mL = 10mg Te (10, 000ppm Te)Te in 10% HNO3 |
Physical Form | Liquid |
Chemical Name or Material | Tellurium ICP Standard |
Safety and Handling
ShelfLife | 18 months |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction