Alfa Aesar™ Tellurium(IV) bromide, ultra dry, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 4343930

Quantity 250g

 View more versions of this product

Catalog No. AA4343930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10031-27-3
    Molecular Formula TeBr4
    Formula Weight 447.27
    MDL Number MFCD00049575
    UN Number UN2923
    Merck Index 14,9125
    Chemical Name or Material Tellurium (IV) bromide, Ultra dry

    Specifications

    Formula Weight 447.27
    Packaging Ampule
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN2923
    Boiling Point 420°C (dec.)
    TSCA TSCA
    Sensitivity Moisture Sensitive
    Melting Point 380°C
    Health Hazard 2 GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 4.31g/mL at 15°C
    Molecular Formula TeBr4
    MDL Number MFCD00049575
    CAS 10031-27-3
    Physical Form Powder
    Merck Index 14,9125
    Quantity 250g
    Health Hazard 1 Danger
    Storage Note 1
    • Ampouled under argon
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Chemical Name or Material Tellurium (IV) bromide, Ultra dry
    Assay 99.999%, (metals basis)

    GHS H Statement
    H300-H314-H318
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Ampouled under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit