Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium(IV) chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
10026-07-0
Cl4Te
269.4
MFCD00011262
SWLJJEFSPJCUBD-UHFFFAOYSA-N
tellurium tetrachloride,telluric chloride,tellurium chloride,tetrachlorotellurium,tellurium iv chloride,tellurium chloride tecl4 , t-4,unii-dny2r5498h,tellurium chloride, t-4,telluricchloride,tecl4
61443
Cl[Te](Cl)(Cl)Cl
10026-07-0
Cl4Te
269.4
MFCD00011262
SWLJJEFSPJCUBD-UHFFFAOYSA-N
tellurium tetrachloride,telluric chloride,tellurium chloride,tetrachlorotellurium,tellurium iv chloride,tellurium chloride tecl4 , t-4,unii-dny2r5498h,tellurium chloride, t-4,telluricchloride,tecl4
61443
Cl[Te](Cl)(Cl)Cl
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Linear Formula TeCl4
Fieser 09,443; 10,377; 12,457; 16,316
Merck Index 15, 9268
Solubility Information Solubility in water: may decompose. Other solubilities: forming teo2 and hcl,soluble in absolute alcohol and toluene
Formula Weight 269.4
Physical Form Powder
Percent Purity 99%
Density 3.2600g/mL
Packaging Glass bottle
Solubility Solubility in water: may decompose. Other solubilities: forming teo2 and hcl, soluble in absolute alcohol and toluene
Specific Gravity 3.26
Boiling Point 380.0°C
Color White to Yellow
Melting Point 224.0°C
Quantity 25g
Chemical Name or Material Tellurium(IV) chloride, 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 233-055-6

RTECSNumber : WY2635000

TSCA : TSCA