Alfa Aesar™ Tellurium(IV) isopropoxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3656504

Quantity 2g

 View more versions of this product

Catalog No. AA3656504

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1795-64-8
    Formula Weight 363.73
    MDL Number MFCD00145354
    UN Number UN1993
    Chemical Name or Material Tellurium(IV) isopropoxide

    Specifications

    Formula Weight 363.73
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    UN Number UN1993
    Health Hazard 1 Warning
    Odor Alcohol-like
    Quantity 2g
    MDL Number MFCD00145354
    Health Hazard 2 GHS H Statement
    H226-H302-H312-H332-H315-H319
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Boiling Point 76°C (0.5mmHg)
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    CAS 1795-64-8
    Sensitivity Moisture & Light Sensitive
    Chemical Name or Material Tellurium(IV) isopropoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H226-H302-H312-H332-H315-H319
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit