Tellurium(IV) oxide, 99.995% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1182130

Quantity 250g

 View more versions of this product

Catalog No. AA1182130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7446-07-3
    Molecular Formula TeO2
    Formula Weight 159.6
    MDL Number MFCD00011263
    Merck Index 14,9123
    Chemical Name or Material Tellurium(IV) oxide, (metals basis)

    Specifications

    Formula Weight 159.6
    Quantity 250g
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    Molecular Formula TeO2
    MDL Number MFCD00011263
    CAS 7446-07-3
    Merck Index 14,9123
    Melting Point 733°C
    Density 5.8
    RTECS Number WY2675000
    Chemical Name or Material Tellurium(IV) oxide, (metals basis)
    Assay 99.995%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit