Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tellurium(IV) oxide, Puratronic, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4782175
Click to view available options
Quantity:
25 g
100 g
500 g
7446-07-3
O2Te
159.60
MFCD00011263
LAJZODKXOMJMPK-UHFFFAOYSA-N
tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq
62638
O=[Te]=O
7446-07-3
O2Te
159.60
MFCD00011263
LAJZODKXOMJMPK-UHFFFAOYSA-N
tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglq
62638
O=[Te]=O

It is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 733°C
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9995%, metal basis
Solubility Information Soluble in strong acids and alkali. Insoluble in water.
IUPAC Name tellurium dioxide
Formula Weight 159.6
Density 5.8
Chemical Name or Material Tellurium(IV) Oxide
Hazard Category H317-H332-H360
Precautionary Statement P201-P202-P261-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-193-1
RTECSNumber WY2675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit