Alfa Aesar™ Tellurium lump, 99.999+% (metals basis)

Manufacturer: Alfa Aesar™ 1036236

Quantity 500g

 View more versions of this product

Catalog No. AA1036236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13494-80-9
    MDL Number MFCD00134062
    Chemical Name or Material Tellurium lump

    Specifications

    Physical Form Lumps
    Quantity 500g
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00134062
    TSCA TSCA
    CAS 13494-80-9
    Health Hazard 1 Danger
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H301-H332
    Toxic if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Tellurium lump
    Assay ≥99.999%, (metals basis)

    GHS H Statement
    H301-H332
    Toxic if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P301+P310-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit